missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitrilotriacetic Acid Disodium Salt, TCI America™

$45.83 - $200.14

Chemical Identifiers

CAS 15467-20-6
Molecular Formula C6H7NNa2O6
Molecular Weight (g/mol) 235.103
MDL Number MFCD00064230
InChI Key RHPXYIKALIRNFA-UHFFFAOYSA-L
Synonym nitrilotriacetic acid disodium salt, kiresuto ntb, disodium nitrilotriacetate, nitrilotriacetic acid, disodium salt, disodium hydrogen nitrilotriacetate, unii-llv4ki749r, disodium nitriloacetate, nitrilotriacetic acid,disodium salt, llv4ki749r, n,n-bis carboxymethyl glycine disodium salt
PubChem CID 27301
IUPAC Name disodium;2-[carboxylatomethyl(carboxymethyl)amino]acetate
SMILES C(C(=O)O)N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N010025G View Documents TCI America
N010025G
25g Each for $45.83
 
N0100500G View Documents TCI America
N0100500G
500g Each for $200.14
 
Specifications

Chemical Identifiers

15467-20-6
235.103
RHPXYIKALIRNFA-UHFFFAOYSA-L
27301
C(C(=O)O)N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+]
C6H7NNa2O6
MFCD00064230
nitrilotriacetic acid disodium salt, kiresuto ntb, disodium nitrilotriacetate, nitrilotriacetic acid, disodium salt, disodium hydrogen nitrilotriacetate, unii-llv4ki749r, disodium nitriloacetate, nitrilotriacetic acid,disodium salt, llv4ki749r, n,n-bis carboxymethyl glycine disodium salt
disodium;2-[carboxylatomethyl(carboxymethyl)amino]acetate

Specifications

White
15467-20-6
MFCD00064230
RHPXYIKALIRNFA-UHFFFAOYSA-L
disodium;2-[carboxylatomethyl(carboxymethyl)amino]acetate
27301
Crystal-Powder at 20°C
Nitrilotriacetic Acid Disodium Salt
25g
C6H7NNa2O6
nitrilotriacetic acid disodium salt, kiresuto ntb, disodium nitrilotriacetate, nitrilotriacetic acid, disodium salt, disodium hydrogen nitrilotriacetate, unii-llv4ki749r, disodium nitriloacetate, nitrilotriacetic acid,disodium salt, llv4ki749r, n,n-bis carboxymethyl glycine disodium salt
C(C(=O)O)N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+]
235.103
235.10
≥99.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1277

RTECSNumber : AJ0350000

TSCA : Yes