Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitrilotriacetic acid, ACS, 98.0% min

Chelating and sequestering agent, used in the manufacture of synthetic detergents

$42.01 - $102.68

Chemical Identifiers

Chemical Name or Material Nitrilotriacetic acid
CAS 139-13-9
Purity Grade ACS Grade
Linear Formula N(CH2CO2H)3
Formula Weight 191.14
MDL Number MFCD00004287
Beilstein 1710776
Merck Index 14,6579
Synonym Tris(carboxymethyl)amine; N,N-Bis(carboxymethyl)glycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3651522

 
alfa aesar™
3651522
100g Each for $42.01

AA3651536

 
alfa aesar™
3651536
500g Each for $102.68
Description & Specifications

Specifications

Formula Weight 191.14
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Nitrilotriacetic acid
TSCA TSCA
Purity Grade ACS Grade
Physical Form Powder
Assay ≤98.0%
Linear Formula N(CH2CO2H)3
MDL Number MFCD00004287
CAS 139-13-9
Beilstein 1710776
Merck Index 14,6579
Synonym Tris(carboxymethyl)amine; N,N-Bis(carboxymethyl)glycine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H302-H315-H319-H335
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number AJ0175000
Odor Odorless
Color White
Melting Point 246°C (dec.)
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H302-H315-H319-H335
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.