missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitrilotriacetic acid, 99%

Manufacturer:  Alfa Aesar™ A1193636

 View more versions of this product

Catalog No. AAA1193636


 

Description

Description

Applications
It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications

Specifications

Nitrilotriacetic acid
246°C (decomposition)
139-13-9
C6H9NO6
MFCD00004287
14,6579
MGFYIUFZLHCRTH-UHFFFAOYSA-N
2-[bis(carboxymethyl)amino]acetic acid
8758
191.14
Odorless
500g
99%
N(CH2CO2H)3
1710776
Tris(carboxymethyl)amine; N,N-Bis(carboxymethyl)glycine
C(C(=O)O)N(CC(=O)O)CC(=O)O
191.139
CHEBI:44557

Chemical Identifiers

139-13-9
191.139
MGFYIUFZLHCRTH-UHFFFAOYSA-N
8758
C6H9NO6
MFCD00004287
Tris(carboxymethyl)amine; N,N-Bis(carboxymethyl)glycine
CHEBI:44557
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302-H315-H319-H335
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AJ0175000

TSCA : TSCA

SDS Documents