Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

$274.18 - $1,379.02

Specifications

Antigen Nitric Oxide Synthase, inducible (iNOS) Ab-1
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1605P

 
thermo scientific™ lab vision™
RB1605P
Purified Ab with BSA and Azide 1mL Each for $1,255.45

RB1605P0

 
thermo scientific™ lab vision™
RB1605P0
Purified Ab with BSA and Azide 100μL Each for $274.18

RB1605P1

 
thermo scientific™ lab vision™
RB1605P1
Purified Ab with BSA and Azide 500μL Each for $754.47

RB1605P1ABX

 
thermo scientific™ lab vision™
RB1605P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $843.24

RB1605PABX

 
thermo scientific™ lab vision™
RB1605PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,379.02
Description & Specifications

Specifications

Antigen Nitric Oxide Synthase, inducible (iNOS) Ab-1
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages
Isotype IgG

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Rat, Mouse and Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages

Molecular Weight: 130kDa

Postitive Control: Rat lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO