Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody


Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1605PABX

 View more versions of this product

Catalog No. RB1605PABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Immunogen A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages
Antigen Nitric Oxide Synthase, inducible (iNOS) Ab-1
Quantity 1mL
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Regulatory Status RUO
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Neuroscience
Concentration 1mg/mL
Conjugate Unlabeled
Type Primary
Classification Polyclonal
Item Description 1mL; 1mg/mL; Unlabeled; Purified without BSA and azide

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Rat, Mouse and Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages

Molecular Weight: 130kDa

Postitive Control: Rat lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO