Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody


Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1605P1ABX

 View more versions of this product

Catalog No. RB1605P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Nitric Oxide Synthase, inducible (iNOS) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages
Isotype IgG
Quantity 500μL
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Human
Target Species Murine

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Nitric Oxide Synthase, inducible (iNOS) Ab-1, Rabbit Polyclonal Antibody.

NOS oxidizes a guanidine nitrogen of arginine releasing nitric oxide in the form of a free radical and citrulline. Nitric oxide thus generated acts as a messenger in diverse functions including vasodilation neurotransmission, anti-tumor and anti-pathogenic activities.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Rat, Mouse and Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of iNOS from activated mouse macrophages

Molecular Weight: 130kDa

Postitive Control: Rat lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO