Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitric acid, fuming, 98%

Chemical Identifiers

Chemical Name or Material Nitric acid
CAS 7697-37-2
Name Note Fuming
Molecular Formula HNO3
Formula Weight 63.01
MDL Number MFCD00011349
UN Number UN2031
Merck Index 14,9840
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45479AN

  SDS
alfa aesar™
45479AN
400mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 63.01
Assay Percent Range 98%
Molecular Formula HNO3
Chemical Name or Material Nitric acid
Sensitivity Hygroscopic
TSCA TSCA
Boiling Point 85°C
Name Note Fuming
Physical Form Liquid
MDL Number MFCD00011349
CAS 7697-37-2
Density 1.5g/mL
Health Hazard 3 GHS P Statement
P221-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Take any precaution to avoid mixing with combustibles.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packaging Group: I; Proper Shipping Name: NITRIC ACID
Color Pale yellow to yellow
Quantity 400mL
pH <1
Melting Point ≈−50°C
Assay 98%
UN Number UN2031
Merck Index 14,9840
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H330-H314-H318
May intensify fire
oxidizer.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Odor Acrid
RTECS Number QU5775000
Safety and Handling

Danger

GHS P Statement
P221-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Take any precaution to avoid mixing with combustibles.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H330-H314-H318
May intensify fire
oxidizer.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.