Learn More
Nitric acid, fuming, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045479AN
Description
Main commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
<1 | |
Yellow | |
∼-50°C | |
85°C | |
Acrid | |
98% | |
HNO3 | |
MFCD00011349 | |
14,9840 | |
Miscible with water and ether. | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
98% |
Nitric acid | |
Fuming | |
7697-37-2 | |
1.5 g/mL | |
Hygroscopic | |
400 mL | |
Liquid | |
UN2031 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 |
Safety and Handling
GHS H Statement
H272-H330-H314-H318
May intensify fire
oxidizer.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H272-H290-H314-H331
DOTInformation : Transport Hazard Class: 8; Packaging Group: I; Proper Shipping Name: NITRIC ACID
EINECSNumber : 231-714-2
RTECSNumber : QU5775000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only