Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitric acid, Environmental Grade Plus, 68-70%

Chemical Identifiers

Chemical Name or Material Nitric acid
CAS 7697-37-2
Purity Grade Environmental Grade Plus
Formula Weight 63.01
MDL Number MFCD00011349
UN Number UN2031
Merck Index 14,6577
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38748AP

  SDS
alfa aesar™
38748AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid, Double Sub-Boiling Quartz Distillation
Formula Weight 63.01
Chemical Name or Material Nitric acid
TSCA TSCA
Boiling Point 121°C
Purity Grade Environmental Grade Plus
Assay 68 to 70%
MDL Number MFCD00011349
CAS 7697-37-2
UN Number UN2031
Merck Index 14,6577
Density 1.41g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.
Health Hazard 3 GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID
Odor Pungent
RTECS Number QU5775000
Color Colorless to pale yellow
Quantity 500mL
Safety and Handling

Danger

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.