Please login to your online account to display your discounted pricing

Nitric acid, Environmental Grade, 67-70%, Alfa Aesar™

$191.36 - $216.97

Chemical Identifiers

Chemical Name or Material Nitric acid
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range 67-70%
Molecular Formula HNO3
Formula Weight 63.01
MDL Number MFCD00011349
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38747AP

  SDS
alfa aesar
38747AP
500mL Each for $191.36

AA38747K4

  SDS
alfa aesar
38747K4
2.5L Each for $216.97
Description & Specifications

Specifications

Formula Weight 63.01
Physical Form Liquid
Quantity 500mL
Color Colorless to pale yellow
DOT Information DOT Class: 8, Packaging Group: II
EINECS Number 231-714-2
Purity Grade Environmental Grade
Molecular Formula HNO3
Vapor Pressure 23 hPa (17mm Hg) at 20°C
CAS 7697-37-2
Odor Pungent
Appearance Colorless to pale yellow liquid
Chemical Name or Material Nitric acid
Concentration or Composition (by Analyte or Components) Nitric acid: 69.0%; Water: 31.0%
CAS 7732-18-5
Health Hazard 1 H314-H272: Causes severe skin burns and eye damage. May intensify fire; oxidizer.
Assay Percent Range 67-70%
MDL Number MFCD00011349
Safety and Handling

H314-H272: Causes severe skin burns and eye damage. May intensify fire; oxidizer.