Please login to your online account to display your discounted pricing

Nitric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

$162.43 - $241.38

Chemical Identifiers

Chemical Name or Material Nitric Acid
CAS 7697-37-2
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Molecular Formula HNO3
Formula Weight 63.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003464

 
avantor performance
936805
Poly Bottle 2.5L Each for $241.38

02003472

 
avantor performance
936801
PFA Bottle 500mL Each for $162.43
Description & Specifications

Specifications

Chemical Name or Material Nitric Acid
Quantity 2.5L
CAS 7697-37-2
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Molecular Formula HNO3
Formula Weight 63.01
Packaging Poly Bottle
Specific Gravity 1L = 1.41kg

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use