missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Nitric acid, ACS Reagent, 70%, Honeywell Fluka™

ACS Reagent, 70%

$113.55 - $154.59

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.012
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym acide nitrique
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES [N+](=O)(O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6001090 View Documents Honeywell-Fluka
438073500ML
500mL Glass Bottle Each for $113.55
 
6001088 View Documents Honeywell Fluka™
43807325L
2.5L Glass Bottle Each for $154.59
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
acide nitrique
CHEBI:48107
[N+](=O)(O)[O-]

Specifications

500mL
≥50.0000%,≥30.0000%
HNO3
MFCD00011349
acide nitrique
[N+](=O)(O)[O-]
1.413 g/mL at 20 °C(lit.)
944
63.01
Glass Bottle
Nitric acid, ACS Reagent, 70%
7697-37-2,7732-18-5
<70.0000%,<50.0000%
HNO3
UN2031
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
63.012
CHEBI:48107
ACS Reagent
122°C
Safety and Handling

Safety and Handling

ShelfLife : 720 days from date of manufacture

DOTInformation : Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Nitric acid

EINECSNumber : 231-714-2

Recommended Storage : Room Temp