Learn More
Nitric Acid, ACS Grade, 68.0% to 70.0% (w/w), LabChem™
Supplier: LabChem LC177003S
Specifications
Nitric Acid | |
7697-37-2 | |
Passes Test | |
Nitrogen oxides; Oxygen | |
HNO3 | |
HNO3 | |
UN2031 | |
Soluble in water | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
68.0 to 70.0% (w/w) |
Colorless | |
100 | |
PVC Coated | |
2.5 L | |
Liquid | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
ACS |
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Keep only in original container.
Do not breathe mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immed
Danger
EINECSNumber : 231-714-2
Recommended Storage : Room Temperature