Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitric acid, ACS, Fuming, 90%, Alfa Aesar™

$164.05 - $164.05

Chemical Identifiers

Chemical Name or Material Nitric acid
CAS 7697-37-2
Purity Grade ACS Grade
Molecular Formula HNO3
Formula Weight 63.01
MDL Number MFCD00011349
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA33261P2

  SDS
alfa aesar
33261P2
1 lb. CALL N/A N/A
 Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Nitric acid
Quantity 1 lb.
CAS 7697-37-2
Health Hazard 1 H314-H272: Causes severe skin burns and eye damage. May intensify fire; oxidizer.
Appearance Colorless to pale yellow liquid
EINECS Number 231-714-2
Purity Grade ACS Grade
Physical Form Liquid
Assay Percent Range 90%
Molecular Formula HNO3
Formula Weight 63.01
MDL Number MFCD00011349
Density 1.41g/cm3 at 20°C
Odor Pungent
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Safety and Handling

H314-H272: Causes severe skin burns and eye damage. May intensify fire; oxidizer.