Please login to your online account to display your discounted pricing

Nitric Acid ACS AR, Macron Fine Chemicals™


macron

Quantity 6 x 2.5L
Packaging Glass S/S

Manufacturer: avantor performance  270448

 View more versions of this product

Catalog No. MK2703212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Nitric Acid
Quantity 6 x 2.5L
CAS 7697-37-2
Purity Grade ACS Grade
Molecular Formula HNO3
Formula Weight 63.01
Packaging Glass S/S

Specifications

Chemical Name or Material Nitric Acid
CAS 7697-37-2
Purity Grade ACS Grade
Molecular Formula HNO3
Formula Weight 63.01