Please login to your online account to display your discounted pricing

Nitric Acid ACS AR, Macron Fine Chemicals™


macron

Quantity 2.5L
Packaging Glass

Manufacturer: avantor performance  270446

 View more versions of this product

Catalog No. MK2704212X6

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 63.01
Purity Grade ACS Grade
Quantity 2.5L
CAS 7697-37-2
Molecular Formula HNO3
Chemical Name or Material Nitric Acid
Packaging Glass

Specifications

Chemical Name or Material Nitric Acid
CAS 7697-37-2
Purity Grade ACS Grade
Molecular Formula HNO3
Formula Weight 63.01