Learn More
Nitric acid, ACS, 68.0-70.0%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 033260R1
Description
Main commercial usage of Nitric acid, ACS, 68.0-70.0% is in the production of fertilizers and reagent for nitration. It is used in the applications for the production of explosives, nylon precursors and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors and in purification processes for raw carbon nanotubes, artificially age pine and maple.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Colorless to pale yellow liquid | |
Colorless to Yellow | |
121°C | |
Pungent | |
68.0-70.0% | |
HNO3 | |
MFCD00011349 | |
Miscible with water and ether. | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
ACS |
Nitric acid | |
7697-37-2 | |
1.41 | |
23 hPa (17mm Hg) at 20°C | |
6 x 7 lb. | |
Liquid | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 |
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Safety and Handling
P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H272-H290-H314-H331
DOTInformation : DOT Class: 8; Packing Group: II
EINECSNumber : 231-714-2
RTECSNumber : QU5775000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only