Please login to your online account to display your discounted pricing

Nitric Acid, 69.0 to 70.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5L
Packaging Glass Bottle

Manufacturer: avantor performance  960105

 View more versions of this product

Catalog No. 02003468

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Assay 69.0 to 70.0%
Formula Weight 63.01
Chemical Name or Material Nitric Acid
Molecular Formula HNO3
CAS 7697-37-2
Specific Gravity 1L = 1.41kg
Packaging Glass Bottle
Quantity 2.5L

Specifications

Chemical Name or Material Nitric Acid
CAS 7697-37-2
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula HNO3
Formula Weight 63.01