Please login to your online account to display your discounted pricing

Nitric Acid, 69.0% to 70.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$130.71 - $140.86

Chemical Identifiers

Chemical Name or Material Nitric Acid
CAS 7697-37-2
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula HNO3
Formula Weight 63.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003467

 
avantor performance
960104
Clear Glass Bottle 2.5L Each for $140.86

02003469

 
avantor performance
960134
Clear PVC Coated Glass Bottle 2.5L Each for $130.71
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Assay 69.0 to 70.0%
Formula Weight 63.01
Chemical Name or Material Nitric Acid
Molecular Formula HNO3
Specific Gravity 1L = 1.41kg
CAS 7697-37-2
Packaging Clear Glass Bottle
Quantity 2.5L