Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitric acid, 65-70%, 99.9999% (metals basis)

Chemical Identifiers

Chemical Name or Material Nitric acid
CAS 7697-37-2
Formula Weight 63.01
MDL Number MFCD00011349
UN Number UN2031
Merck Index 14,6577
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10984AC

 
alfa aesar™
10984AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10984AK

 
alfa aesar™
10984AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10984AP

 
alfa aesar™
10984AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 63.01
Chemical Name or Material Nitric acid
TSCA TSCA
Boiling Point 121°C
Physical Form Liquid
Assay 99.9999%
Assay Percent Notes (metals basis)
MDL Number MFCD00011349
CAS 7697-37-2
UN Number UN2031
Merck Index 14,6577
Concentration 65 to 70%
Density 1.41g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.
Health Hazard 3 GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID
Odor Pungent
RTECS Number QU5775000
Color Colorless to pale yellow
Quantity 25mL
Safety and Handling

Danger

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.