missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitric acid, 65-70%, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 10984AK

 View more versions of this product

Catalog No. AA10984AK



Specifications

Specifications

Nitric acid
250mL
(metals basis)
MFCD00011349
14,6577
GRYLNZFGIOXLOG-UHFFFAOYSA-N
65 to 70%
1.41g/mL
944
63.01
99.9999%
121°C
Colorless to Yellow
7697-37-2
HNO3
UN2031
acide nitrique
[N+](=O)(O)[O-]
nitric acid
63.012
CHEBI:48107
Liquid
Pungent

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
acide nitrique
CHEBI:48107
[N+](=O)(O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID

RTECSNumber : QU5775000

TSCA : TSCA

Documents