Learn More
Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals
For Use in the Manufacture of Fertilizers and Explosives, in organic Synthesis, in Metallurgy
$98.81 - $207.99
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Description
In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuelsMain commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Specifications
Colorless to Yellow | |
121°C | |
Pungent | |
HNO3 | |
MFCD00011349 | |
14,6577 | |
Very soluble in water and ether; Decomposes in alcohol;Miscible with water and ether. | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
99.999% | |
(metals basis) |
7697-37-2 | |
1.41 | |
100 mL | |
Liquid | |
UN2031 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
Nitric acid, 65 to 70% |
Safety and Handling
GHS H Statement
H314-H272;
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H272-H290-H314-H331
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID
EINECSNumber : 231-714-2
RTECSNumber : QU5775000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only