missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Nitric Acid, 6.00 Normal, Ricca Chemical
Chemical Identifiers
CAS |
7697-37-2 |
Molecular Formula |
HNO3 |
Molecular Weight (g/mol) |
63.012 |
InChI Key |
GRYLNZFGIOXLOG-UHFFFAOYSA-N |
Synonym |
acide nitrique, acidum nitricum, aqua fortis, azotic acid, hydrogen nitrate, nital, nitrous fumes, nitryl hydroxide, rfna, salpetersaeure |
PubChem CID |
944 |
ChEBI |
CHEBI:48107 |
IUPAC Name |
nitric acid |
SMILES |
[N+](=O)(O)[O-] |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
54-405-PT
View Documents
|
Ricca Chemical Company
RI54405PT
|
2.5L |
Glass clear |
Each for $658.82
|
|
Add to cart
|
|
|
Description
Specifications
Chemical Identifiers
|
7697-37-2 |
|
63.012 |
|
acide nitrique, acidum nitricum, aqua fortis, azotic acid, hydrogen nitrate, nital, nitrous fumes, nitryl hydroxide, rfna, salpetersaeure |
|
CHEBI:48107 |
|
[N+](=O)(O)[O-] |
|
HNO3 |
|
GRYLNZFGIOXLOG-UHFFFAOYSA-N |
|
944 |
|
nitric acid |
Specifications
|
2.5L |
|
6.000 ± 0.002 N at 20°C |
|
acide nitrique, acidum nitricum, aqua fortis, azotic acid, hydrogen nitrate, nital, nitrous fumes, nitryl hydroxide, rfna, salpetersaeure |
|
[N+](=O)(O)[O-] |
|
63.012 |
|
CHEBI:48107 |
|
Glass clear |
|
Aqueous Solution |
|
7697-37-2,7732-18-5 |
|
HNO3 |
|
GRYLNZFGIOXLOG-UHFFFAOYSA-N |
|
nitric acid |
|
944 |
|
Slightly pungent to pungent |
|
Solubility in Water: Infinite |
|
Nitric Acid, 6.00 Normal |
SDS