missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitric acid, 2N Standardized Solution, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 44528K2

 View more versions of this product

Catalog No. AA44528K2



Specifications

Specifications

Nitric Acid, 2N Standardized Solution
7697-37-2
MFCD00011349
Miscible with water and ether.
[N+](=O)(O)[O-]
1.06 g/mL
944
63.01
1L
HNO3
acide nitrique
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
63.012
CHEBI:48107
Liquid

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
acide nitrique
CHEBI:48107
[N+](=O)(O)[O-]
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H332

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents