missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nitric Acid, Certified, 10.0% (v/v) ±0.5%, LabChem™
Supplier: LabChem LC177302
Specifications
Nitric Acid (1+9) | |
7697-37-2,7732-18-5 | |
90.5,9.5 | |
Amber Glass | |
Traceable to NIST | |
1 L | |
Liquid | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
63.01 | |
Certified |
Colorless | |
1.05g/mL | |
Passes Test | |
1.05g/mL | |
Nitrogen oxides; Thermal decomposition generates: Corrosive vapors | |
HNO3 | |
HNO3 | |
UN2031 | |
Soluble in water | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
10.0% ±0.5% (v/v) |
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
944 | |
nitric acid |
HNO3 | |
MFCD00011349 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 | |
O[N+]([O-])=O |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse m
Danger
Recommended Storage : Room Temperature