Please login to your online account to display your discounted pricing

Nitrendipine, MP Biomedicals™


Quantity 5mg

Manufacturer: mp biomedicals  0215978905

 View more versions of this product

Catalog No. ICN15978905

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5mg
Chemical Name or Material Nitrendipine
CAS 39562-70-4
Physical Form Lemon Yellow Powder
Molecular Formula C18H20N2O6
Formula Weight 360.37g/mol

Specifications

Chemical Name or Material Nitrendipine
CAS 39562-70-4
Physical Form Lemon Yellow Powder
Molecular Formula C18H20N2O6
Formula Weight 360.37g/mol