Please login to your online account to display your discounted pricing

Nitrendipine, MP Biomedicals™

$54.92 - $54.92

Chemical Identifiers

Chemical Name or Material Nitrendipine
CAS 39562-70-4
Physical Form Lemon Yellow Powder
Molecular Formula C18H20N2O6
Formula Weight 360.37g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15978905

 
mp biomedicals
0215978905
5mg Each for $54.92
Description & Specifications

Specifications

Quantity 5mg
Chemical Name or Material Nitrendipine
CAS 39562-70-4
Physical Form Lemon Yellow Powder
Molecular Formula C18H20N2O6
Formula Weight 360.37g/mol