Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitrated Peptide Certified Mass Spec Standard Mixture II

For cellular and molecular biology applications

Specifications

Product Type Nitrated Peptide Certified Mass Spec Standard Mixture II, 500pmol each peptide per vial
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Nitrated ACTH (1-10), Nitrated Angiotensin I, Nitrated SCPA, and non-nitrated partners ACTH (1-10), Angiotensin I, SCPA
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67809LPL

 
alfa aesar™
J67809LPL
1 vial, 500 pmol CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67809ODL

 
alfa aesar™
J67809ODL
3 vials, 500 pmol/vial CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Nitrated Peptide Certified Mass Spec Standard Mixture II, 500pmol each peptide per vial
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Nitrated ACTH (1-10), Nitrated Angiotensin I, Nitrated SCPA, and non-nitrated partners ACTH (1-10), Angiotensin I, SCPA
Regulatory Status RUO
Purification Method HPLC