Please login to your online account to display your discounted pricing

Alfa Aesar™ NitRated Angiotensin I, Certified Mass Spec grade

For research use

Manufacturer: alfa aesar™  J67672-LPL

 View all options for this product

Catalog No. AAJ67672LPL

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications


Format Lyophilized powder
Formulation Lyophilized powder
Quantity 1 vial
Recombinant No
Storage Requirements -30° to -10°C
Regulatory Status RUO
Protein Length DRV-Y(NO2)-IHPFHL
Product Type NitRated Angiotensin I, Certified Mass Spec grade