missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
LaMotte™ Nitrate-Nitrogen Standard Solution
Used with LaMotte SMART™ Spectro Spectrophometer.
Supplier: LaMotte™ 5392H
Description
- Size: 2 oz. (60mL)
- Concentration: 1000ppm
- Non-hazardous
Specifications
LaMotte SMART™ Spectro Spectrophometer | |
Nitrate-Nitrogen Standard solution | |
60 mL |