Please login to your online account to display your discounted pricing

LaMotte™ Nitrate-Nitrogen Kit

Colorimetric test system.

$80.70 - $80.70

Specifications

Range 1 to 15ppm
Includes 50 nitrate No. 1 tablets, 50 nitrate No. 2 test tablets, two plastic test tubes with caps, Octa slide viewer with slide bar, instructions
View More Specs
Includes:  50 Nitrate No. 1 tablets, 50 Nitrate No. 2 test tablets, two plastic test tubes with caps, Octa-Slide viewer with slide bar, instructions
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S65410

 
lamotte™
3354
Nitrate Nitrogen Tablet Test Kit Each for $80.70
Description & Specifications

Specifications

Range 1 to 15ppm
Includes 50 nitrate No. 1 tablets, 50 nitrate No. 2 test tablets, two plastic test tubes with caps, Octa slide viewer with slide bar, instructions

  • Range: 0 to 15ppm
  • Sensitivity: 1.0ppm
  • Number of Tests: 50