missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nitrate (as Nitrogen) Standard, 1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 4.43mg NO3-), Certified, LabChem™
Supplier: LabChem LC179101
Specifications
Nitrate (as Nitrogen) Standard | |
1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 4.43mg NO3-) | |
Poly Bottle | |
99.08,0.72,0.198,0.002 | |
as N | |
KNO3 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
potassium nitrate | |
24434 | |
101.1 | |
Passes Test | |
Liquid |
1000ppm ±10ppm (1mL = 1mg N, 1mL = 4.43mg NO3-) | |
7757-79-1,67-66-3,64-17-5,7732-18-5 | |
500 mL | |
Colorless | |
KNO3 | |
MFCD00011409 | |
[K+].[O-][N+]([O-])=O | |
101.10 | |
CHEBI:63043 | |
Certified | |
Soluble in water |
Chemical Identifiers
7757-79-1 | |
101.10 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
CHEBI:63043 | |
[K+].[O-][N+]([O-])=O |
KNO3 | |
MFCD00011409 | |
24434 | |
potassium nitrate |
Safety and Handling
GHS H Statement
May cause cancer (oral, inhalation).
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature