missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitinol Wire, 0.5mm (0.02 in.) dia., Alfa Aesar™

Form: 13.07g/10m; Af Approx. 10deg.C; Superelastic; Oxide surface

Chemical Identifiers

Physical Form Wire, 0.5mm (0.02 in.) dia.
MDL Number MFCD02091734
Chemical Name or Material Nitinol, Alloy N, Straight Annealed
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA44950G6 SDS Alfa Aesar
44950G6
10m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA44950H2 SDS Alfa Aesar
44950H2
50m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA44950H5 SDS Alfa Aesar
44950H5
150m N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

Wire, 0.5mm (0.02 in.) dia.
MFCD02091734
Nitinol, Alloy N, Straight Annealed

Specifications

Silver-gray
Silver-gray wire
Wire, 0.5mm (0.02 in.) dia.
Nitinol, Alloy N, Straight Annealed
10m
Odorless
MFCD02091734

Safety and Handling

H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.