missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitinol wire, , 0.58mmAlfa Aesar™

Form: approx. 17.29g/10m; Af approx. 10°C; Superelastic; Oxide surface

Manufacturer:  Alfa Aesar 44951H5

 View more versions of this product

Catalog No. AA44951H5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Nitinol
150m
Silver-gray wire
MFCD02091734
Silver to Gray
Alloy N, straight annealed
Odorless
Wire, 0.58mm (0.023 in.) dia.
Documents