missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitinol Wire, 0.41mm (0.016 in.) dia., Alfa Aesar™

Form: Approx. 8.37g/10m; Af Approx. 10 deg.C; Superelastic; Oxide surface

Manufacturer:  Alfa Aesar 44949G6

 View more versions of this product

Catalog No. AA44949G6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Nitinol
10m
Silver-gray wire
MFCD02091734
Silver to Gray
Alloy N, Straight Annealed
Odorless
Wire, 0.41mm (0.016 in.) dia.
Documents