Please login to your online account to display your discounted pricing

Nitinol tension coil spring, Alloy M, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Nitinol tension coil spring
Name Note Wire diameter 0.75mm, mean coil diameter 5.55mm
CAS 7440-02-0
CAS 7440-32-6
Physical Form Solid
MDL Number MFCD02091734
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45655KJ

  SDS
alfa aesar
45655KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA45655KM

  SDS
alfa aesar
45655KM
25 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA45655KN

  SDS
alfa aesar
45655KN
100 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solid
Color Silvery gray
Quantity 5 pieces
Chemical Name or Material Nitinol tension coil spring
Odor Odorless
Name Note Wire diameter 0.75mm, mean coil diameter 5.55mm
MDL Number MFCD02091734
CAS 7440-02-0
Concentration or Composition (by Analyte or Components) Nickel: 55.75%; Titanium: 44.25%
Health Hazard 1 H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
CAS 7440-32-6
Safety and Handling

H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.