Alfa Aesar™ Nitinol foil, 0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface
Manufacturer:  Alfa Aesar™ 44952FI
 View more versions of this product
Catalog No. AA44952FI
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Nitinol foil
50 x 50mm
Odorless
Af approx. 10°C
≈0.51g/25x25mm
0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface
MFCD02091734
Documents