Alfa Aesar™ Nitinol foil, 0.05mm (0.002 in.) thick, superelastic, flat annealed, pickled surface
Manufacturer:  Alfa Aesar™ 45492FF
 View more versions of this product
Catalog No. AA45492FF
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Nitinol foil
25 x 25mm
Odorless
Af approx. 10°C
≈0.25g/25x25mm
0.05mm (0.002 in.) thick, superelastic, flat annealed, pickled surface
MFCD02091734
Documents