Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitinol foil, 0.025mm (0.001 in.) thick, shape memory, flat annealed, pickled surface

Chemical Identifiers

Chemical Name or Material Nitinol foil
Name Note 0.025mm (0.001 in.) thick, shape memory, flat annealed, pickled surface
MDL Number MFCD02091734
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47169FF

  SDS
alfa aesar™
47169FF
25 x 25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47169FI

  SDS
alfa aesar™
47169FI
50 x 50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47169GE

  SDS
alfa aesar™
47169GE
75 x 100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25 x 25mm
MDL Number MFCD02091734
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Chemical Name or Material Nitinol foil
Name Note 0.025mm (0.001 in.) thick, shape memory, flat annealed, pickled surface
Odor Odorless
Weight ≈0.1024g/25x25mm
TSCA TSCA
Physical Form Af approx. 45°C
Health Hazard 3 GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.