Alfa Aesar™ Nitinol compression coil spring, wire diameter 1.0mm (0.039in), mean coil diameter 7.0mm (0.28in), Alloy M
Manufacturer:  Alfa Aesar™ 45951KM
 View more versions of this product
Catalog No. AA45951KM
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Nitinol compression coil spring, Alloy M
Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.), mean coil diameter 7.0mm (0.28 in.)
MFCD02091734
25 pc.
Odorless
Af approx. 41°C
Documents