Please login to your online account to display your discounted pricing

Nital Etchant, 10%, Ricca Chemical

$378.48 - $845.47

Chemical Identifiers

Chemical Name or Material Nital Etchant
CAS 64-17-5
CAS Min % 73.0
CAS Max % 76.0
CAS 7697-37-2
CAS Min % 16.92
CAS Max % 17.61
CAS 67-63-0
CAS Min % 3.87
CAS Max % 4.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R53230004A

 
ricca chemical company
RIR53230004A
Poly natural 4L Each for $845.47

R5323000250

 
ricca chemical company
RIR5323000250A
Poly natural 250mL Each for $378.48
Description & Specifications

Specifications

Color Clear colorless to pale-yellow
Physical Form Liquid
Quantity 4L
Concentration or Composition Notes 10%
CAS 67-63-0
CAS 64-17-5
Packaging Poly natural
CAS Max % 17.61
CAS 67-63-0
Chemical Name or Material Nital Etchant
CAS Max % 4.03
CAS Min % 73.0
CAS Max % 76.0
CAS Min % 16.92
CAS Min % 3.87
CAS 7697-37-2