Please login to your online account to display your discounted pricing

Alfa Aesar™ NIST traceable size standard, polystyrene monodisperse, 500nm, 1% wt%, aqueous suspension in dropper-tipped bottles

Chemical Identifiers

Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 500nm
CAS 9003-53-6
Name Note 1% wt%, aqueous suspension
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ677084Y

  SDS
alfa aesar™
J677084Y
15mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 500nm
CAS 9003-53-6
Physical Form Liquid
Density 1.05-g/mL
Storage Note 1 2° to 8°C
Color White
Quantity 15mL
TSCA TSCA
Name Note 1% wt%, aqueous suspension
Odor Odorless
Packaging dropper-tipped bottles
Safety and Handling

2° to 8°C