missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ NIST traceable size standard, polystyrene monodisperse, 3μm, 1% wt%, aqueous suspension in dropper-tipped bottles

Manufacturer:  Alfa Aesar™ J677584Y

 View more versions of this product

Catalog No. AAJ677584Y

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

NIST traceable size standard, polystyrene monodisperse, 3μm
9003-53-6
Liquid
dropper-tipped bottles
15mL
1% wt%, aqueous suspension
1.050g/mL
Odorless
White

Chemical Identifiers

9003-53-6
NIST traceable size standard, polystyrene monodisperse, 3μm, 1% wt%, aqueous suspension

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

storageNote1 : 2° to 8°C