missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ NIST traceable size standard, polystyrene monodisperse, 20μm, 1% wt%, aqueous suspension in dropper-tipped bottles

Chemical Identifiers

CAS 9003-53-6
Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 20μm, 1% wt%, aqueous suspension
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ677794Y SDS Alfa Aesar™
J677794Y
15mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

9003-53-6
NIST traceable size standard, polystyrene monodisperse, 20μm, 1% wt%, aqueous suspension

Specifications

9003-53-6
White
15mL
dropper-tipped bottles
Liquid
Odorless
1.050g/mL
NIST traceable size standard, polystyrene monodisperse, 20μm, 1% wt%, aqueous suspension

Safety and Handling

TSCA : TSCA

storageNote1 : 2° to 8°C