Please login to your online account to display your discounted pricing

Alfa Aesar™ NIST traceable size standard, polystyrene monodisperse, 100nm, 1% wt%, aqueous suspension in dropper-tipped bottles

$486.42 - $486.42

Chemical Identifiers

Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 100nm
CAS 9003-53-6
Name Note 1% wt%, aqueous suspension
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ675324Y

 
alfa aesar™
J675324Y
15mL Each for $486.42
Description & Specifications

Specifications

CAS 9003-53-6
TSCA TSCA
Name Note 1% wt%, aqueous suspension
Physical Form Liquid
Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 100nm
Density 1.050g/mL
Odor Odorless
Packaging dropper-tipped bottles
Storage Note 1 2° to 8°C
Color White
Quantity 15mL
Safety and Handling

2° to 8°C