Please login to your online account to display your discounted pricing

Alfa Aesar™ NIST traceable size standard, polystyrene monodisperse, 1.5μm, 1% wt%, aqueous suspension in dropper-tipped bottles


Quantity 15mL

Manufacturer: alfa aesar™  J676174Y

 View more versions of this product

Catalog No. AAJ676174Y

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 1.5μm
CAS 9003-53-6
Physical Form Liquid
Name Note 1% wt%, aqueous suspension
Storage Note 1 2° to 8°C
Packaging dropper-tipped bottles
Color White
Quantity 15mL
Density 1.050g/mL
Odor Odorless
TSCA TSCA

Specifications

Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 1.5μm
CAS 9003-53-6
Name Note 1% wt%, aqueous suspension
Accessories

2° to 8°C

SDS & Safety Documents