Please login to your online account to display your discounted pricing

Alfa Aesar™ NIST traceable size standard, polystyrene monodisperse, 1.5μm, 1% wt%, aqueous suspension in dropper-tipped bottles

Chemical Identifiers

Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 1.5μm
CAS 9003-53-6
Name Note 1% wt%, aqueous suspension
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ676174Y

  SDS
alfa aesar™
J676174Y
15mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material NIST traceable size standard, polystyrene monodisperse, 1.5μm
CAS 9003-53-6
Physical Form Liquid
Name Note 1% wt%, aqueous suspension
Storage Note 1 2° to 8°C
Packaging dropper-tipped bottles
Color White
Quantity 15mL
Density 1.050g/mL
Odor Odorless
TSCA TSCA
Safety and Handling

2° to 8°C