Please login to your online account to display your discounted pricing

Nisoxetine hydrochloride, Tocris


Noradrenalin re-uptake inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS102510

 View more versions of this product

Catalog No. 102510

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 307.82
Quantity 10mg
Chemical Name or Material Nisoxetine hydrochloride
Assay Percent Range >99%
Molecular Formula C17H21NO2·HCl
CAS 57754-86-6
Synonym (±)-γ-(2-Methoxyphenoxy)-N-methylbenzenepropanamine hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Nisoxetine hydrochloride
Assay Percent Range >99%
Molecular Formula C17H21NO2·HCl
Formula Weight 307.82
Synonym (±)-γ-(2-Methoxyphenoxy)-N-methylbenzenepropanamine hydrochloride
Accessories

Store at Room Temperature