Please login to your online account to display your discounted pricing

Nisoxetine hydrochloride, Tocris

Noradrenalin re-uptake inhibitor

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material Nisoxetine hydrochloride
Assay Percent Range >99%
Molecular Formula C17H21NO2·HCl
Formula Weight 307.82
Synonym (±)-γ-(2-Methoxyphenoxy)-N-methylbenzenepropanamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

102510

 
r & d systems
RDS102510
10mg Each for $146.74

102550

 
r & d systems
RDS102550
50mg Each for $618.87
Description & Specifications

Specifications

Chemical Name or Material Nisoxetine hydrochloride
Quantity 10mg
CAS 57754-86-6
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C17H21NO2·HCl
Formula Weight 307.82
Synonym (±)-γ-(2-Methoxyphenoxy)-N-methylbenzenepropanamine hydrochloride
Safety and Handling

Store at Room Temperature