Learn More
Nipecotic acid, 98%, Thermo Scientific Chemicals
$59.89 - $374.97
Chemical Identifiers
CAS | 498-95-3 |
---|---|
Molecular Formula | C6H11NO2 |
Molecular Weight (g/mol) | 129.159 |
MDL Number | MFCD00005992 |
InChI Key | XJLSEXAGTJCILF-UHFFFAOYSA-N |
Synonym | nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine |
PubChem CID | 4498 |
ChEBI | CHEBI:116931 |
IUPAC Name | piperidine-3-carboxylic acid |
SMILES | C1CC(CNC1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2472303
|
Thermo Scientific Chemicals
B2472303 |
1 g |
Each for $59.89
|
|
|||||
AAB2472306
|
Thermo Scientific Chemicals
B2472306 |
5 g |
Each for $108.95
|
|
|||||
AAB2472314
|
Thermo Scientific Chemicals
B2472314 |
25 g |
Each for $374.97
|
|
|||||
Description
GABA reuptake inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
498-95-3 | |
129.159 | |
XJLSEXAGTJCILF-UHFFFAOYSA-N | |
4498 | |
piperidine-3-carboxylic acid |
C6H11NO2 | |
MFCD00005992 | |
nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine | |
CHEBI:116931 | |
C1CC(CNC1)C(=O)O |
Specifications
498-95-3 | |
C6H11NO2 | |
1 g | |
14,6561 | |
Soluble in water | |
C1CC(CNC1)C(=O)O | |
129.159 | |
CHEBI:116931 | |
98% |
∼261°C (decomposition) | |
MFCD00005992 | |
81096 | |
nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine | |
XJLSEXAGTJCILF-UHFFFAOYSA-N | |
piperidine-3-carboxylic acid | |
4498 | |
129.16 | |
Nipecotic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-873-9
RTECSNumber : TM6125380
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only